Orders LinkContactUsLinkComponent
Safety spectacles, Virtua™ AP
Safety spectacles, Virtua™ AP
Anti-UV, anti-scratch, grå, Grey PC
Katalog # MMMAVIRTUAA1
Leverandør:   3M
Safety spectacles, Virtua™ AP
Katalog # MMMAVIRTUAA1
Leverandør:  3M
Safety spectacles, Virtua™ AP
Safety spectacles, Virtua™ AP
Anti-UV, anti-scratch, grå, Grey PC
Price
38,40 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Lightweight, stylish designed safety glasses Virtua™ AP protective eyewear features a streamline wrap-around design for excellent coverage and good field of vision.
  • Integrated brow bar for added protection
  • Lightweight for maximum comfort
  • Strong polycarbonate lenses featuring anti-scratch coating for improved vision and extended durability
Weight: 26 g
 : EN 166:2001
Specifikationer
  • Model:
    Anti-UV, anti-scratch
  • Farve:
    grå
  • Seq:
    02
  • Linser:
    Grey PC