BASIC Plus Laboratory Coats
blå, M
Katalog # KIMB10031
Gå til:
Discontinued
Product Details & Documents
These coats are made of a protective three-layer SMS fabric which is resistant to blood strike-through. Recommended for areas where there is the potential for light fluid contact.
- Knit collar and cuffs
- Five snap buttons
- Three reinforced pockets
- With size/name label
- Neck loop for hanging
Specifikationer
- Tøj størrelse:M
- Farve:blå
- Seq:2
- Køn:Unisex
- Cat. No.:KIMB10031
- Outfit applikationer:Laboratory|Clinical
- Lukkestørrelse:Med trykknapper
- Environmentally Preferable:Reduktion af affald
- Lommer:med lommer