Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Carbon standards

Carbon standards

Inorganic carbon standard, 5 ppm, 500 mL
  REGETIC5
 :   REAGECON
Carbon standards
  REGETIC5
 :  REAGECON
 :  
Carbon standards

Inorganic carbon standard, 5 ppm, 500 mL

 :  
 
2.840,00 kr
   
1 * 500 mL
 

These standards are manufactured from high purity raw materials and is suitable for calibrating a wide range of Total Organic Carbon (TOC) and Total Inorganic Carbon (TIC) analysers to a high accuracy.

These standards are independent, traceable and certified.

Certifications: Accredited to IEC/ISO 17025 and ISO 17034.

Documents

 

 

  • Koncentration:
    5 ppm
  • Pakn:
    500 mL
  • Beskrivelse:
    Inorganic carbon standard