Compensating Cables for Temperature Sensors
Compensating cable, type J, sort/hvid, 10 m
Katalog # WINKWFZJ0250
Leverandør: WINKLER
Gå til:
Product Details & Documents
Compensating cables in PTFE insulated, twisted leads.
Specifikationer
- Farve:sort/hvid
- Beskrivelse:Compensating cable, type J
- Længde:10 m
- Seq:01