Temperature sensors
Thermocouple tape, type J (NiCr-Ni), with wire end ferrules and cable (1,2 m), +500…+1000 °C, 16×450 mm
Katalog # WINKWFBK1999-XX012
Leverandør: WINKLER
Gå til:
Product Details & Documents
Specifikationer
- Område:+500…+1000 °C
- Beskrivelse:Thermocouple tape, type J (NiCr-Ni), with wire end ferrules and cable (1,2 m)
- Seq:04
- Ø×L:16×450 mm