Heating tapes
Heating tape, 16,0 m length, 1570 W
Katalog # WINKWBS00216001570
Leverandør: WINKLER
Gå til:
Product Details & Documents
These heating tapes contain four heat conductors, insulated silicone, outer braiding with tinned copper wire, factory terminated and vulcanised main connection.
- Very high and constant heating power per meter
- High operating temperature
- Extremely flexible
- Quick and easy installation
Specifikationer
- Beskrivelse:Heating tape, 16,0 m length, 1570 W
- Seq:24