Double spatulas, micro
130 mm, 40 mm, 2 mm
Catalog # WITG7.412.051
Supplier: witeg Labortechnik
Jump to:
Product Details & Documents
Micro spatulas with two flat ends, made of stainless steel.
Specifications
- Længde:130 mm
- Bladbredde:2 mm
- Klinge længde:40 mm