Counting chambers, Malassez
Counting chamber with single ruling, 0,20 mm
Katalog # HECH40453701
Leverandør: GLASWARENFABRIK KARL HECHT
Gå til:
Product Details & Documents
These counting chambers are designed for blood tests and urine analysis.
Specifikationer
- Kammerdybde:0,20 mm
- Beskrivelse:Counting chamber with single ruling
- Seq:002