Orders LinkContactUsLinkComponent
Accessories for test sieves
Accessories for test sieves
Intermediate pan, 300 mm, 60 mm
Katalog # HAVE205923777
Leverandør:   NEXOPART
Accessories for test sieves
Katalog # HAVE205923777
Leverandør:  NEXOPART
Accessories for test sieves
Accessories for test sieves
Intermediate pan, 300 mm, 60 mm
Price
1.400,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Ø:
    300 mm
  • Beskrivelse:
    Intermediate pan
  • Højde:
    60 mm
  • Seq:
    10