Test panels
Test panel, harmonic steel, smooth, degreased and cleaned, 120×200×0,15 mm
Katalog # SIMEVF8515
Leverandør: Industrial Physics
Gå til:
Product Details & Documents
Test panels are used to enhance reproducibility of physical and chemical tests. Each panel is equipped with a hole for hanging and handling.
- Consistent and reproducible quality with sophisticated finishing: No burrs or sharp edges
- Physical testing: Adhesion, elongation, impact resistance and elasticity
- Appearance testing of coatings (gloss, colour, texture)
Documents
TQC Company Video
Videos
Specifikationer
- Beskrivelse:Test panel, harmonic steel, smooth, degreased and cleaned
- Seq:21
- B×D×H:120×200×0,15 mm