Orders LinkContactUsLinkComponent
Accessories for adhesion test kits
Accessories for adhesion test kits
Nylon brush for cross cut adhesion test
Catalog # SIMESP1710
Supplier:   Industrial Physics
Accessories for adhesion test kits
Catalog # SIMESP1710
Supplier:  Industrial Physics
Accessories for adhesion test kits
Accessories for adhesion test kits
Nylon brush for cross cut adhesion test
Price
138,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Beskrivelse:
    Nylon brush for cross cut adhesion test
  • Seq:
    11
Accessories
Product Information
  • Catalog #
    Beskrivelse
    Bredde
    Availability
    Price
  • SIMESP3010
    Tape for cross cut adhesion test, 3 rolls of 11,0 m, acc. to ISO 2409:2003
    25,0 mm
    Log in for availability
    1 * 3 ROL - 285,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Bredde
    Availability
    Price
  • SIMESP3007
    Tape for cross cut adhesion test, 1 roll of 11,0 m, acc. to ISO 2409:2003
    25,0 mm
    Log in for availability
    1 * 1 Items - 193,00 kr