Orders LinkContactUsLinkComponent
Mandrels for cylindrical bend testers
Mandrels for cylindrical bend testers
Set of 14 mandrels (Ø = 2, 3, 4, 5, 6, 8, 10, 12, 13, 16, 19, 20, 25, 32 mm) with desktop holder
Katalog # SIMESP1835
Leverandør:   Industrial Physics
Mandrels for cylindrical bend testers
Katalog # SIMESP1835
Leverandør:  Industrial Physics
Mandrels for cylindrical bend testers
Mandrels for cylindrical bend testers
Set of 14 mandrels (Ø = 2, 3, 4, 5, 6, 8, 10, 12, 13, 16, 19, 20, 25, 32 mm) with desktop holder
Price
4.090,00 kr
Log ind for tilgængelighed
1 * 14 Items
Product Details & Documents
Documents
Specifikationer
  • Beskrivelse:
    Set of 14 mandrels (Ø = 2, 3, 4, 5, 6, 8, 10, 12, 13, 16, 19, 20, 25, 32 mm) with desktop holder
  • Seq:
    923