Accessories for adhesion test kits
Spare cutting blade, 11 teeths, 1 mm, acc. to ISO 2409, suitable for CC2000 and CC3000 test kits
Catalog # SIMESP1705
Supplier: Industrial Physics
Jump to:
Product Details & Documents
Specifications
- Beskrivelse:Spare cutting blade, 11 teeths, 1 mm, acc. to ISO 2409, suitable for CC2000 and CC3000 test kits
- Seq:02
Accessories
Product Information
- Catalog #BeskrivelseBreddeAvailabilityPrice
- SIMESP3010Tape for cross cut adhesion test, 3 rolls of 11,0 m, acc. to ISO 2409:200325,0 mmLog in for availability1 * 3 ROL - 285,00 kr
Product Information
- Catalog #BeskrivelseBreddeAvailabilityPrice
- SIMESP3007Tape for cross cut adhesion test, 1 roll of 11,0 m, acc. to ISO 2409:200325,0 mmLog in for availability1 * 1 Items - 193,00 kr