Accessories for adhesion test kits
Spare cutting blade, 11 teeths, 1,5 mm, acc. to ASTM D3359 <2009, suitable for CC2000 and CC3000 test kits
Katalog # SIMESP1706
Leverandør: Industrial Physics
Gå til:
Product Details & Documents
Documents
TQC Company Video
Videoer
Specifikationer
- Beskrivelse:Spare cutting blade, 11 teeths, 1,5 mm, acc. to ASTM D3359 <2009, suitable for CC2000 and CC3000 test kits
Accessories
Produktinformation
- Katalog #BreddeTilgængelighedPrice
- Tape for cross cut adhesion test, 3 rolls of 11,0 m, acc. to ISO 2409:2003Tape for cross cut adhesion test, 3 rolls of 11,0 m, acc. to ISO 2409:2003SIMESP301025,0 mmLog ind for tilgængelighed1 * 3 ROL - 285,00 kr
Produktinformation
- Katalog #BreddeTilgængelighedPrice
- Tape for cross cut adhesion test, 1 roll of 11,0 m, acc. to ISO 2409:2003Tape for cross cut adhesion test, 1 roll of 11,0 m, acc. to ISO 2409:2003SIMESP300725,0 mmLog ind for tilgængelighed1 * 1 Items - 193,00 kr