Orders LinkContactUsLinkComponent
Film applicators, multi-functional
Film applicators, multi-functional
Multi-functional film applicator, 4 sided, custom gaps, width 80 mm
Katalog # SIMEVF2172
Leverandør:   Industrial Physics
Film applicators, multi-functional
Katalog # SIMEVF2172
Leverandør:  Industrial Physics
Film applicators, multi-functional
Film applicators, multi-functional
Multi-functional film applicator, 4 sided, custom gaps, width 80 mm
Price
5.610,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Multi-functional film applicator with 4 application sides for applying paint films of 4 different predefined thicknesses. The applicators are available in film widths of 60, 80 mm and one side of the applicator is supplied with a guidance support for straight application. Made from high grade stainless steel, the multi-functional film applicator will not be affected by acid or base elements.
Documents
Specifikationer
  • Beskrivelse:
    Multi-functional film applicator, 4 sided, custom gaps, width 80 mm
  • Seq:
    05