Surface profile comparators
Surface profile comparator for sand
Catalog # SIMELD2051
Supplier: Industrial Physics
Jump to:
Product Details & Documents
Surface comparators for determining roughness by touch and sight.
- Five different grades of roughness in each comparator
- Designed to be used with the illuminated magnifier
- Available in Imperial units only
Section profiles: 0,5 ; 1,0 ; 2,0 ; 3,0 ; 4,0 mils for the three versions.
Certifications: ASTM D4417
Documents
TQC Company Video
Videos
Specifications
- Beskrivelse:Surface profile comparator for sand
Accessories
Product Information
- Catalog #BeskrivelseAvailabilityPrice
- SIMELD2054Surface profile disc holder and magnifier (5×)Log in for availability1 * 1 Items - 929,00 kr