Calibration standards for thickness gauges
Coating thickness metal calibration plate, 0; 75; 250; 1500 µm
Katalog # 649-0020
Leverandør: Industrial Physics
Gå til:
Product Details & Documents
These coated metal calibration standards consist of a set of four calibration plates.
They are composed of an epoxy coating on steel or aluminium when many gauges are in use or when accuracy verification is performed frequently.
Certifications: ISO/QS-9000 and in-house quality control requirements.
Documents
TQC Company Video
Videos
Specifikationer
- Område:0; 75; 250; 1500 µm
- Beskrivelse:Coating thickness metal calibration plate
- Seq:01