Orders LinkContactUsLinkComponent
Calibration standards for thickness gauges
Calibration standards for thickness gauges
Coating thickness calibration standard, 0; 15; 40; 100 µm
Catalog # 649-0022
Supplier:   Industrial Physics
Calibration standards for thickness gauges
Catalog # 649-0022
Supplier:  Industrial Physics
Calibration standards for thickness gauges
Calibration standards for thickness gauges
Coating thickness calibration standard, 0; 15; 40; 100 µm
Price
4.250,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
These coated metal calibration standards consist of a set of four calibration plates.
They are composed of an epoxy coating on steel or aluminium when many gauges are in use or when accuracy verification is performed frequently.
Certifications: ISO/QS-9000 and in-house quality control requirements.
Documents
Specifications
  • Område:
    0; 15; 40; 100 µm
  • Beskrivelse:
    Coating thickness calibration standard
  • Seq:
    03