Calibration standards for thickness gauges
Coating thickness calibration standard, 0; 15; 40; 100 µm
Catalog # 649-0022
Supplier: Industrial Physics
Jump to:
Product Details & Documents
These coated metal calibration standards consist of a set of four calibration plates.
They are composed of an epoxy coating on steel or aluminium when many gauges are in use or when accuracy verification is performed frequently.
Certifications: ISO/QS-9000 and in-house quality control requirements.
Documents
TQC Company Video
Videos
Specifications
- Område:0; 15; 40; 100 µm
- Beskrivelse:Coating thickness calibration standard
- Seq:03