Orders LinkContactUsLinkComponent
Coating thickness testers, PosiTector® 6000
Coating thickness testers, PosiTector® 6000
Microprobe, N90S, 0 - 625 µm (±0,5 +1 % (0 - 100) ; ±2 +3 % (over 100)) for non conductive layers on NFe metals
Catalog # SIMELD6108
Supplier:   Industrial Physics
Coating thickness testers, PosiTector® 6000
Catalog # SIMELD6108
Supplier:  Industrial Physics
Coating thickness testers, PosiTector® 6000
Coating thickness testers, PosiTector® 6000
Microprobe, N90S, 0 - 625 µm (±0,5 +1 % (0 - 100) ; ±2 +3 % (over 100)) for non conductive layers on NFe metals
Price
kr7,860.00
Log in for availability
1 * 1 Items
Product Details & Documents
This coating thickness gauge is a fully electronic apparatus, that uses magnetic and eddy current principles, to measure coating thickness on both ferrous (Fe) and non ferrous (NFe) metals.
  • Fast measurement for quick inspection
  • Flip display enables right side up viewing
  • USB port for fast connection to a PC with continuous power
Ordering information: Software Ideal Finish Analysis is available on request. With two user levels Ideal Finish Analysis offers user friendly reporting functions for standard production work, as well as advanced calculations for in depth analysis. For the complete range of available probes, please contact your local Avantor agency. Please order probes separately.
Delivery information: Supplied with plastic shims, rubber holster with belt clip and wrist strap, three batteries, instructions, nylon carrying case with shoulder strap, certificate of calibration traceable to NIST and USB cable.
Documents
Specifications
  • Model:
    N90S
  • Område:
    0 - 625 µm (±0,5 +1 % (0 - 100) ; ±2 +3 % (over 100)) for non conductive layers on NFe metals
  • Beskrivelse:
    Microprobe
  • Seq:
    126
Accessories
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • SIMELD5505
    Infrared printer
    Log in for availability
    1 * 1 Items - 3.270,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • SIMELD6015
    USB cable
    Log in for availability
    1 * 1 Items - 903,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • SIMELD5508
    Bluetooth printer
    Log in for availability
    1 * 1 Items - 4.560,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Område
    Availability
    Price
  • 649-0022
    Coating thickness calibration standard
    0; 15; 40; 100 µm
    Log in for availability
    1 * 1 Items - 4.250,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Område
    Availability
    Price
  • 649-0023
    Coating thickness calibration standard
    0; 75; 250; 1500 µm
    Log in for availability
    1 * 1 Items - 4.250,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Område
    Availability
    Price
  • 649-0025
    Coating thickness calibration standard
    75; 125; 250; 500 µm
    Log in for availability
    1 * 1 Items - 4.250,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • SIMELD6016
    PosiTector smartlink, Defelsko
    Log in for availability
    1 * 1 Items - 5.380,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • SIMELD5407
    CAL P1 standard
    Log in for availability
    1 * 1 Items - 2.360,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • SIMELD5408
    CAL P2 standard
    Log in for availability
    1 * 1 Items - 2.360,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • SIMELD5409
    CAL P3 standard
    Log in for availability
    1 * 1 Items - 2.360,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • SIMELD5410
    CAL P4 standard
    Log in for availability
    1 * 1 Items - 2.360,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Model
    Område
    Availability
    Price
  • SIMELD6017
    90° angle probe
    FRS
    0 - 1500 µm (±1 +1 % (0 - 50) ; ±2 +1 % (over 50)) for NFe metals
    Log in for availability
    1 * 1 Items - 5.380,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Model
    Område
    Availability
    Price
  • SIMELD6025
    Probe
    F
    0 - 1500 µm (±1 +1 % (0 - 50) ; ±2 +1 % (over 50)) for NFe metals
    Log in for availability
    1 * 1 Items - 3.830,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Model
    Område
    Availability
    Price
  • SIMELD6020
    Probe
    FKS
    0 - 13 000 mm (±0,02 +3 % (0 - 13)) for thick NFe metals
    Log in for availability
    1 * 1 Items - 6.530,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Model
    Område
    Availability
    Price
  • SIMELD6023
    Built-in detachable probe
    FNTS
    0 - 6000 mm (±0,01 +1 % (0 - 2,5) ; ±0,01 +3 % (over 2,5 mm)) for all metals
    Log in for availability
    1 * 1 Items - 9.780,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Model
    Område
    Availability
    Price
  • SIMELD6022
    90° angle probe
    FNRS
    0 - 1500 µm (±1 +1 % (0 - 50) ; ±2 +1 % (over 50)) for all metals
    Log in for availability
    1 * 1 Items - 9.210,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Model
    Område
    Availability
    Price
  • SIMELD6018
    Built-in detachable probe
    FTS
    0 - 6000 mm (±0,01 +1 % (0 - 2,5) ; ±0,01 +3 % (over 2,5)) for non-magnetic coatings over ferrous metals (steel and iron)
    Log in for availability
    1 * 1 Items - 6.010,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Model
    Område
    Availability
    Price
  • SIMELD6024
    Probe
    NKS
    0 - 13 000 mm (±0,02 +3 % (0 - 13)) for non conductive NFe metals
    Log in for availability
    1 * 1 Items - 6.730,00 kr