Lab Markers, several colours
Lab marker, fine tip, sort
Katalog # PRCI1400-20-PDC
Marketsource
Leverandør: PRECISION DYNAMICS
Gå til:
Product Details & Documents
Versatile, permanent markings write on plastic, metal, and porcelain labware.
- Several colours
- Unique ink has the ability to write on labware even when the surface is damp
Specifikationer
- Farve:sort
- Beskrivelse:Lab marker, fine tip
- Seq:02