Filtering apparatus, DURAN®
Complete with 1000 ml filtration bottle, 250 ml filter head, PTFE insert and clamp
Catalog # 516-0431
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
Filtration apparatus with unit made from clear DURAN® borosilicate glass 3.3, PTFE insert and anodised aluminium clamp.
- Scaled funnel
- All components are available as spare parts
Typical applications: Coarse and ultrafine filtration, filtration of HPLC media, residue analysis. Filter paper, membrane filters (47 mm) or glass filters can be used for filtration.
Documents
257105451
Images 360
Specifications
- Beskrivelse:Complete with 1000 ml filtration bottle, 250 ml filter head, PTFE insert and clamp
- Seq:01
Accessories
Product Information
- Catalog #BeskrivelseAvailabilityPrice
Product Information
- Catalog #BeskrivelseAvailabilityPrice
Product Information
- Catalog #BeskrivelseAvailabilityPrice
Product Information
- Catalog #BeskrivelseMaterialeUdv.ØAvailabilityPrice
- 201-2101Hose connection, straight, with silicone seal (VMQ)PP8.6 mmLog in for availability1 * 1 Items - 15,30 kr
Product Information
- Catalog #BeskrivelseGevindFarveAvailabilityPrice
- 201-1921Screw cap with aperture, height: 28 mm, bore: 34 mmGL 14rødLog in for availability1 * 1 Items - 17,20 kr
Product Information
- Catalog #BeskrivelseAvailabilityPrice
Product Information
- Catalog #BeskrivelseAvailabilityPrice
Product Information
- Catalog #BeskrivelseAvailabilityPrice
Product Information
- Catalog #BeskrivelseAvailabilityPrice
Product Information
- Catalog #BeskrivelseAvailabilityPrice