Accessories
Multi-purpose adapter, MSAA502, for eliminating ambient light or providing an enhanced field of view at optimal distances, compatible with Dino-Lite AM models (excluding models with polariser)
Katalog # IDCPMSAA502
Leverandør:   Dino-Lite / IDCP B.V.
Accessories
Katalog # IDCPMSAA502
Leverandør:  Dino-Lite / IDCP B.V.
Accessories
Accessories
Multi-purpose adapter, MSAA502, for eliminating ambient light or providing an enhanced field of view at optimal distances, compatible with Dino-Lite AM models (excluding models with polariser)
Price
181,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Multi-purpose adapter, MSAA502, for eliminating ambient light or providing an enhanced field of view at optimal distances, compatible with Dino-Lite AM models (excluding models with polariser)