Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Threaded fittings

Threaded fittings

Tubing, bend with cap nut, nozzle-Ø 10 mm, NW 20 mm, PP, M36×3
Katalog # BURK8548-0025
Leverandør:   BURKLE
Threaded fittings
Katalog # BURK8548-0025
Leverandør:  BURKLE
Threaded fittings
Threaded fittings

Tubing, bend with cap nut, nozzle-Ø 10 mm, NW 20 mm, PP, M36×3

Price
58,80 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Gevind:
    M36×3
  • Beskrivelse:
    Tubing, bend with cap nut, nozzle-Ø 10 mm, NW 20 mm
  • Materiale:
    PP