HPLC columns, Acclaim™ Trinity™ P1
Analytisk, 3 µm, 2,10 mm, 50 mm
Katalog # 555-7461
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
The Acclaim™ Trinity™ P1 is a unique, high-efficiency, silica-based column designed for pharmaceutical applications, such as simultaneous separation of pharmaceutical drug substances and their counterions.
- Adjustable selectivity by changing mobile phase ionic strength, electrolyte type, pH and organic solvent
- Ideal selectivity for simultaneous separation of basic, neutral, and acidic analytes
- Retention of ionic and ionisable analytes without using ion-pairing reagents
Particle type: ultra pure, spherical silica
Max. pressure: 4500 psi
Flow rate range: 0,30 to 1,60 ml/min for 3,0 mm ID formats; 0,15 to 0,80 ml/min for 2,1 mm ID formats
Bonded phase: reversed-phase and anion-exchange
pH range: 2,5 to 7,0
Specifikationer
- Kolonne form:Analytisk
- Ø indv.:2,10 mm
- Partikelstørrelse:3 µm
- Længde:50 mm
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Stainless steel guard cartridge holder, V-2Stainless steel guard cartridge holder, V-2554-4528Log ind for tilgængelighed1 * 1 Items - 3.610,00 kr
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



