HPLC columns, Acclaim™ Trinity™ P1
Beskyttelsespatron, 3 µm, 2,10 mm, 10 mm
Katalog # 555-7211
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
The Acclaim™ Trinity™ P1 is a unique, high-efficiency, silica-based column designed for pharmaceutical applications, such as simultaneous separation of pharmaceutical drug substances and their counterions.
- Adjustable selectivity by changing mobile phase ionic strength, electrolyte type, pH and organic solvent
- Ideal selectivity for simultaneous separation of basic, neutral, and acidic analytes
- Retention of ionic and ionisable analytes without using ion-pairing reagents
Particle type: ultra pure, spherical silica
Max. pressure: 4500 psi
Flow rate range: 0,30 to 1,60 ml/min for 3,0 mm ID formats; 0,15 to 0,80 ml/min for 2,1 mm ID formats
Bonded phase: reversed-phase and anion-exchange
pH range: 2,5 to 7,0
Specifikationer
- Kolonne form:Beskyttelsespatron
- Ø indv.:2,10 mm
- Partikelstørrelse:3 µm
- Længde:10 mm
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Stainless steel guard cartridge holder, V-2Stainless steel guard cartridge holder, V-2554-4528Log ind for tilgængelighed1 * 1 Items - 3.610,00 kr



