Warning signs
Hazard label, corrosive substances, 'Vorsicht! Ätzende Stoffe', PP, 262×371 mm
Katalog # 116-0691
Gå til:
Discontinued
Product Details & Documents
These safety signs are for the purposes of accident prevention, fire protection, health hazard information and emergency evacuation.
- Recommended by ISO 7010
- Colour: Yellow/Black
- Shape: Triangle
Specifikationer
- Beskrivelse:Hazard label, corrosive substances, 'Vorsicht! Ätzende Stoffe'
- Dimensioner:262×371 mm
- Materiale:PP
- Seq:29