Orders LinkContactUsLinkComponent
Adapter kit, Neuros™
Adapter kit, Neuros™
Adapter kit for 700/1700 series Neuros™ syringes, 3, 33
Katalog # HAMI65460-01
Leverandør:   HAMILTON BONADUZ
Adapter kit, Neuros™
Katalog # HAMI65460-01
Leverandør:  HAMILTON BONADUZ
Adapter kit, Neuros™
Adapter kit for 700/1700 series Neuros™ syringes, 3, 33
Price
2.810,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
The Neuros™ adapter kit attaches to any 5,0 to 100 µl removable needle (RN) syringe.
  • Gas sterilisable, not autoclavable
  • Stainless steel, borosilicate glass fluid path
The needle sleeves come in two versions, one with a blind stop and one without. The sleeve with a stop is designed for applications where it is desirable to limit the penetration depth of the needle, as is the case with a cannulated animal. The sleeve without a stop supports the needle but does not obstruct the view of the injection target which is ideal for microsurgeries using a stereotaxic holder.
Leveringsinformation: The kit includes a glass adapter, two needle sleeves and a six pack of 33 gauge needles.
Specifikationer
  • Måler:
    33
  • Spidstype:
    3
  • Beskrivelse:
    Adapter kit for 700/1700 series Neuros™ syringes
  • Seq:
    01