Adapter kit, Neuros™
Product Details & Documents
The Neuros™ adapter kit attaches to any 5,0 to 100 µl removable needle (RN) syringe.
- Gas sterilisable, not autoclavable
- Stainless steel, borosilicate glass fluid path
The needle sleeves come in two versions, one with a blind stop and one without. The sleeve with a stop is designed for applications where it is desirable to limit the penetration depth of the needle, as is the case with a cannulated animal. The sleeve without a stop supports the needle but does not obstruct the view of the injection target which is ideal for microsurgeries using a stereotaxic holder.
Leveringsinformation: The kit includes a glass adapter, two needle sleeves and a six pack of 33 gauge needles.
Specifikationer
- Måler:33
- Spidstype:3
- Beskrivelse:Adapter kit for 700/1700 series Neuros™ syringes
- Seq:01