Orders LinkContactUsLinkComponent
Handheld digital microscopes, Dino-Lite Special Light, UV
UV microscope, AM4113T-VW, 10 - 70×, 200×, 4 + 4 LEDs (390/400 nm UV + white), 1,3 MP
  IDCPAM4113T-VW
Handheld digital microscopes, Dino-Lite Special Light, UV
  IDCPAM4113T-VW
Handheld digital microscopes, Dino-Lite Special Light, UV
Handheld digital microscopes, Dino-Lite Special Light, UV
UV microscope, AM4113T-VW, 10 - 70×, 200×, 4 + 4 LEDs (390/400 nm UV + white), 1,3 MP
kr3,610.00
1 * 1 Items
These handheld microscopes feature ultraviolet (UV) light or a combination between UV and white light.
  • High speed USB 2.0 output
  • Metal housing or composite housing; metal models are ESD-safe
  • Edge series models with high-quality optics provide a very sharp, bright and natural colour image with very low aberration and vignetting
  • Wireless streaming to a tablet, smartphone, or computer is possible in combination with WF-10 unit (to be ordered separately)
  • Integrated polariser greatly reduces glare on reflective surfaces (some models only)
  • Exchangeable caps for more flexibility (some models only)
 : Supplied with carry pouch, software CD (DinoCapture 2.0 (Windows), DinoXcope (Mac OS)), user manual, Quick guide and calibration target.
  • Forstørrelse:
    10 - 70×, 200×
  • Model:
    AM4113T-VW
  • Opløsning:
    1,3 MP
  • Beskrivelse:
    UV microscope
  • Seq:
    08
  • Belysning:
    4 + 4 LEDs (390/400 nm UV + white)
Accessories
  •  
    Beskrivelse
    B×D×H
  • IDCPRK-06A
    Tabletop stand for handheld Dino-Lite microscopes, RK-06-A, incl. extension arm RK-10-EX
    150×220×270 mm
    1 * 1 Items - 1.880,00 kr
  •  
    Beskrivelse
  • 630-2923
    Wi-Fi streamer with operating manual, power adapter and 1880 Ah Li-Ion battery
    1 * 1 Items - 2.460,00 kr
  •  
    Beskrivelse
  • 630-1821
    Foot pedal, enables users to take a picture with their foot
    1 * 1 Items - 472,00 kr
  •  
    Beskrivelse
  • 630-2517
    Sturdy high-end stand for Dino-Lite microscopes, RK-10A, incl. HD-P1 holder and RK-10-EX horizontal arm
    1 * 1 Items - 2.540,00 kr