Methylenblåt 10 g/l in aqueous solution
Glass bottle, 1 L
Leverandør: VWR Chemicals
Methylenblåt 10 g/l in aqueous solution
Katalog # 6129.1000
Leverandør: VWR Chemicals
CAS nummer: 61-73-4
Gå til:
Product Details & Documents
Synonymer: Basic Blue 9, Methylthioninium chloride, Methylene blue chloride, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride, Swiss Blue
- CAS nummer:61-73-4
- Formel:C₁₆H₁₈ClN₃S
- MDL nummer:MFCD00150008
- Molecular Weight:319.86 g/mol
- Tæthed:1,01 g/cm³ (20 °C)
- EINECS:200-515-2
Documents
Certifikater
Søg efter partinummerSpecifikationer
Specifikationstestresultater
- Identification:Passes test
- Control weight:QC checked
Specifikationer
- Pakning:Glass bottle
- Pakn:1 L
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



