Transfection reagent, Turbofect™
TurboFect™ transfection reagent, 5×200 µl, 5
Katalog # PIERR0534
Leverandør: Thermo Fisher Scientific
Transfection reagent, Turbofect™
Katalog # PIERR0534
Leverandør: Thermo Fisher Scientific
CAS nummer:
Gå til:
Product Details & Documents
TurboFect™ reagent an easy to use, non-immunogenic transfection reagent for delivery of DNA plasmids and expression vectors into eukaryotic cells.
- High transfection efficiency of a wide variety of cell types
- No optimisation required for most cell lines
- Excellent transfection efficiency in the presence or absence of serum
- Minimal cytotoxicity
- Ready to use without any need to reconstitute, dilute or manipulate
TurboFect™ reagent is ideal for stable and transient transfection or co-transfection of DNA or RNA, experiments with adherent or suspension cells, differentiated or undifferentiated cells and gene expression studies and assays involving GFP or luciferase reporters.
Specifikationer
Specifikationer
- Pakn:5
- Beskrivelse:TurboFect™ transfection reagent, 5×200 µl