Signs, warning stands
Do not enter, black and red on yellow, -, 300×500 mm
Katalog # BRDY256712
Gå til:
Discontinued
Product Details & Documents
These stable warning devices are easy to transport and suitable for the marking of hazardous zones.
- Made of high-density polypropylene
- Durable, yellow
- No buckling or corrosion
- Large note field is visible from over 9 meters
- With default notices or unlabeled also available
Height: 660 mm
Specifikationer
- Beskrivelse:Do not enter, black and red on yellow
- Dimensioner:300×500 mm
- Materiale:-
- Seq:05
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



