Orders LinkContactUsLinkComponent
VWR®, Separating funnels, conical
50 ml, 19/26 NS
Katalog # 532-0012
Leverandør:   VWR Collection
VWR®, Separating funnels, conical
Katalog # 532-0012
Leverandør:  VWR Collection
VWR®, Separating funnels, conical
VWR®, Separating funnels, conical
50 ml, 19/26 NS
Price
389,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Borosilicate 3.3 glass, with NS-PE stopper.
  • With or without graduations
  • Stopcock with PTFE key
  • Very good chemical and temperature resistance
Specifikationer
  • Kappestr.:
    19/26 NS
  • Kapacitet:
    50 ml
  • Seq:
    07