Orders LinkContactUsLinkComponent
VWR®, Separating funnels, conical
VWR®, Separating funnels, conical
2000 ml, 29/32 NS
Katalog # 532-0011
Leverandør:   VWR Collection
VWR®, Separating funnels, conical
Katalog # 532-0011
Leverandør:  VWR Collection
VWR®, Separating funnels, conical
VWR®, Separating funnels, conical
2000 ml, 29/32 NS
Price
1.110,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Borosilicate 3.3 glass, with NS-PE stopper.
  • With or without graduations
  • Stopcock with PTFE key
  • Very good chemical and temperature resistance
Specifikationer
  • Kappestr.:
    29/32 NS
  • Kapacitet:
    2000 ml
  • Seq:
    06