Phanerogames prepared slides
Epidermis of Allium cepa (onion), w.m. showing simple plant cells with cell walls, nuclei and cytoplasm
Katalog # LIEDAS111C
Leverandør: LIEDER JOHANNES
Gå til:
Product Details & Documents
These seagrass preparations are available sets of five.
Specifikationer
- Beskrivelse:Epidermis of Allium cepa (onion), w.m. showing simple plant cells with cell walls, nuclei and cytoplasm
- Seq:02