Orders LinkContactUsLinkComponent
Erlenmeyer flasks, with Quickfit® ground joint
Erlenmeyer flasks, with Quickfit® ground joint
100 ml, 109 mm, 64 mm, 19/26 NS
Katalog # BIBB1132/06
Leverandør:   SciLabware
Erlenmeyer flasks, with Quickfit® ground joint
Katalog # BIBB1132/06
Leverandør:  SciLabware
Erlenmeyer flasks, with Quickfit® ground joint
Erlenmeyer flasks, with Quickfit® ground joint
100 ml, 109 mm, 64 mm, 19/26 NS
Price
2.550,00 kr
Log ind for tilgængelighed
1 * 20 Items
Product Details & Documents
Erlenmeyer flasks, made of Pyrex® borosilicate glass, with standard ground joint.
  • Resistance to thermal shock and chemical corrosion
  • White enamel graduations and marking spot
  • Erlenmeyer flasks with high quality Quickfit® ground glass socket
Specifikationer
  • Udv.Ø:
    64 mm
  • Kappestr.:
    19/26 NS
  • Kapacitet:
    100 ml
  • Højde:
    109 mm
  • Seq:
    02