Objektglas med fordybning
Microscope slides with reaction wells, blå, 3, 14 mm
MARI1216491
Cavity slides for diagnosis with well wettable reaction wells and frosted marking area of 20 mm at one end.
- Soda-lime glass with epoxy resin colour masks
- High resistance against solvents
Documents
- Ant. brønde:3
- Farve:blå
- Beskrivelse:Microscope slides with reaction wells
- Seq:10
- Brønd Ø:14 mm