Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Aprons, plastic, Selefa®

Aprons, plastic, Selefa®

transparent, 810×1250 mm, Roll in dispenser box
Katalog # 113-2012
Leverandør:   Onemed Sverige AB
Aprons, plastic, Selefa®
Katalog # 113-2012
Leverandør:  Onemed Sverige AB
Aprons, plastic, Selefa®
Aprons, plastic, Selefa®

transparent, 810×1250 mm, Roll in dispenser box

Price
136,00 kr
Log ind for tilgængelighed
1 * 50 Items
Product Details & Documents

These disposable aprons are suitable for use in medical areas, such as hospitals and nursing stations.

  • Protect clothing from spills and splashes during medical procedures or cleaning tasks
  • Support infection control

Colour: White or transparent

Emballering: Either packed in bags or in dispenser boxes.

Specifikationer
  • Pakket:
    Roll in dispenser box
  • Farve:
    transparent
  • Seq:
    3
  • B×L:
    810×1250 mm
  • Cat. No.:
    113-2012
  • Outfit applikationer:
    Medical|Clinical
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.