Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Aprons, plastic, Selefa®

Aprons, plastic, Selefa®

transparent, 810×1250 mm, In dispenser box
Katalog # 113-1999
Leverandør:   Onemed Sverige AB
Aprons, plastic, Selefa®
Katalog # 113-1999
Leverandør:  Onemed Sverige AB
Aprons, plastic, Selefa®
Aprons, plastic, Selefa®

transparent, 810×1250 mm, In dispenser box

Price
135,00 kr
Log ind for tilgængelighed
1 * 50 Items
Product Details & Documents

These disposable aprons are suitable for use in medical areas, such as hospitals and nursing stations.

  • Protect clothing from spills and splashes during medical procedures or cleaning tasks
  • Support infection control

Colour: White or transparent

Emballering: Either packed in bags or in dispenser boxes.

Specifikationer
  • Pakket:
    In dispenser box
  • Farve:
    transparent
  • Seq:
    2
  • B×L:
    810×1250 mm
  • Cat. No.:
    113-1999
  • Outfit applikationer:
    Medical|Clinical