Orders LinkContactUsLinkComponent
Stoppers, hollow, rubber
8 mm, 19 mm, 14 mm, 21 mm
  DENE2152021
Stoppers, hollow, rubber
  DENE2152021
Stoppers, hollow, rubber
Stoppers, hollow, rubber
8 mm, 19 mm, 14 mm, 21 mm
1.440,00 kr
1 * 100 Items
  • Ø bund:
    14 mm
  • Ø top:
    21 mm
  • Hul Ø:
    8 mm
  • Højde:
    19 mm
  • Seq:
    5