Orders LinkContactUsLinkComponent
Titrators, SIMPLEX
Titrators, SIMPLEX
For cream, 0 - 40° SH
Katalog # GERB4521
Leverandør:   GERBER FUNKE
Titrators, SIMPLEX
Katalog # GERB4521
Leverandør:  GERBER FUNKE
Titrators, SIMPLEX
Titrators, SIMPLEX
For cream, 0 - 40° SH
Price
1.740,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These SIMPLEX titrators consist of a polyethylene bottle on a plastic base.
  • Burette with automatic zero point adjustment
  • Precision titration by button control
Used for milk and cream.
Leveringsinformation: Supplied with one 1 ml and one 25 ml pipette, one 200 ml Erlenmeyer flask.
Specifikationer
  • Beskrivelse:
    For cream, 0 - 40° SH
  • Seq:
    02