Øjenskyllevæske
Strap for wall bracket, for extra support for the bottle when located in vibrating environments
Katalog # 118-0248
Leverandør: Orkla Care AB / Cederroth
Gå til:
Product Details & Documents
Sterile, buffered, isotonic sodium chloride solution for rinsing eyes following an accident. Available as a single-use, pocket version (235 ml) or wall mounted (500 ml). The bottle opens automatically when twisted out of the wall bracket, and must therefore, be used immediately.
- Quick and easy to open
- Eye cup protected from dust
- Generous flow of fluid for effective rinsing - the eye cup on the bottle channels the fluid to the eye, avoiding unnecessary spills
- Neutralises the pH value
Holdbarhed ved uåbnet flaske er 4 år.
* Vægholder er leveret uden øjenskylleflaske.
Documents
Specifikationer
- Beskrivelse:Strap for wall bracket, for extra support for the bottle when located in vibrating environments
- Seq:04