Accessories
Inlay filter, incl. spring ring for funnels Ø 120 mm, 119×2 mm, 119-0145, -0132, -0133, -0149, -0131, -0153, -2410, -0134, -0151, -0152
Katalog # 119-0166
Leverandør: ROETZMEIER SICHERH BEHAELTER
Gå til:
Specifikationer
- Diameter×H:119×2 mm
- Til:119-0145, -0132, -0133, -0149, -0131, -0153, -2410, -0134, -0151, -0152
- Beskrivelse:Inlay filter, incl. spring ring for funnels Ø 120 mm
- Seq:65