Accessories
Rotable spout 1½“, 28×175 mm, All Rötzmeier safety canisters and barrels (10 and 25 l)
Katalog # 119-0161
Leverandør: ROETZMEIER SICHERH BEHAELTER
Gå til:
Specifikationer
- Diameter×H:28×175 mm
- Til:All Rötzmeier safety canisters and barrels (10 and 25 l)
- Beskrivelse:Rotable spout 1½“
- Seq:41