Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Harnesses, Titan™

Harnesses, Titan™

Harness, Polyamide, 0,5 kg
Katalog # 139-0232
Leverandør:   HONEYWELL SAFETY
Harnesses, Titan™
Katalog # 139-0232
Leverandør:  HONEYWELL SAFETY
Harnesses, Titan™
Harnesses, Titan™

Harness, Polyamide, 0,5 kg

Price
461,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Non stretch harnesses for construction work. Available as a 1-point version with rear anchorage or as a 2-point version with front and rear anchorage.

  • Lightweight, durable polyester webbing for all-day comfort and dependability
  • Fully adjustable designs so one size fits all
  • Sliding back D-ring for a comfortable fit
  • Sub pelvic strap keeps the wearer upright
  • Chest strap (45 mm) with metal buckles

EN 361

Certifications: EN 361

Specifikationer
  • Beskrivelse:
    Harness, Polyamide
  • Vægt:
    0,5 kg
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.