Fleece jacket, DYNAMIC® Tortolas 443
mørkeblå, S
SIOE443Z/B95/S
: SIOEN
Single-coloured jacket for men, made of 2-layer bonded polyester fleece. Both sides are brushed, one side features an anti-pilling treatment. Can be zipped into different jackets.
- Straight collar, hidden zip closure
- Inset chest pocket with zip closure, two inset pockets with zip closure
- Round inset sleeves, elastic piping at end of sleeves
- Drawstring in hem
Weight: 320 g/m²
: EN 340 (1XXXX)
- Tøj størrelse:S
- Farve:mørkeblå
- Seq:01
- Køn:For mænd
- Cat. No.:SIOE443Z/B95/S
- Sleeve or leg length:Long
- Risk area:Cold
- Vægt:320 g/m²
- Model:Tortolas 443
- Outfit applikationer:General Purpose
- Lukkestørrelse:Med lynlås
- Stof:Polyester