Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Bump caps, Bump Cap™ S19

Bump caps, Bump Cap™ S19

Bump Cap™ for the food industry, rød
Catalog # 111-0962
Bump caps, Bump Cap™ S19
Catalog # 111-0962
Bump caps, Bump Cap™ S19
Bump caps, Bump Cap™ S19

Bump Cap™ for the food industry, rød

Price
74,40 kr
Log in for availability
1 * 1 Items
Product Details & Documents

This polyethylene cap is ideal for use in the food industry and in other areas where textile clothing materials are not allowed for reasons of hygiene and where sterility is required.

  • With three point plastic inner cradle
  • Sweatband made ​​from PVC, LDPE headband and interiors are washable and replaceable
  • Lightweight and durable
  • Outer shell is easily washable

Size: 54 to 59 cm

Weight: 195 g

Certifications: BS EN 812: 1997; amendment no. 1 (2001) with CE marking

Specifications
  • Farve:
    rød
  • Beskrivelse:
    Bump Cap™ for the food industry
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.