Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Safety helmets, Concept Roofer

Safety helmet, unvented, with ratchet, EU, grøn
  111-0864
Safety helmets, Concept Roofer
  111-0864
Safety helmets, Concept Roofer
Safety helmets, Concept Roofer

Safety helmet, unvented, with ratchet, EU, grøn

 
 
   
1 * 1 Items
 

The Concept Roofer is a light, strong and comfortable helmet made from ABS.

  • With premium ratchet headband ensuring secure fit
  • Six point terylene webbing cradle
  • Hydro-flock sweatband

Size: 520 to 630 mm

Weight: 340 g

EN 397

 : EN 397

 
  • Farve:
    grøn
  • Beskrivelse:
    Safety helmet, unvented, with ratchet, EU
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.