Orders LinkContactUsLinkComponent
Racks for BOD bottles, WHEATON®
Racks for BOD bottles, WHEATON®
Rack, for 20 BOD bottles (60 ml)
Katalog # WHEAW227729
Leverandør:   DWK Life Sciences
Racks for BOD bottles, WHEATON®
Katalog # WHEAW227729
Leverandør:  DWK Life Sciences
Racks for BOD bottles, WHEATON®
Racks for BOD bottles, WHEATON®
Rack, for 20 BOD bottles (60 ml)
Price
1.560,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Wire, coated.
  • Good chemical resistance
  • Good temperature resistance
These stackable racks transports BOD bottles conveniently and safely.
Specifikationer
  • Beskrivelse:
    Rack, for 20 BOD bottles (60 ml)
  • Seq:
    01