Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Threaded fittings

Tubing nozzle, fix, bend, external thread, nozzle-Ø 12 mm, NW 7,5 mm, PP, 12,7 mm
Catalog # BURK8547-0012
Supplier:   BURKLE
Threaded fittings
Catalog # BURK8547-0012
Supplier:  BURKLE
Threaded fittings
Threaded fittings

Tubing nozzle, fix, bend, external thread, nozzle-Ø 12 mm, NW 7,5 mm, PP, 12,7 mm

Price
30,50 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Gevind:
    12,7 mm
  • Beskrivelse:
    Tubing nozzle, fix, bend, external thread, nozzle-Ø 12 mm, NW 7,5 mm
  • Materiale:
    PP