Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Cortisol express ELISA kit

Cortisol express ELISA kit

Cortisol Express ELISA kit, 480 strip wells, 480 Tests
Katalog # CAYM500370-480
Leverandør:   Cayman Chemical
Cortisol express ELISA kit
Katalog # CAYM500370-480
Leverandør:  Cayman Chemical
CAS nummer:  
Cortisol express ELISA kit
Cortisol express ELISA kit

Cortisol Express ELISA kit, 480 strip wells, 480 Tests

CAS nummer:  
Price
13.530,00 kr
Log ind for tilgængelighed
1 * 0,5 mL
Product Details & Documents

Cortisol Express ELISA kit offers a fast, convenient assay format for measurement of cortisol.

  • Measure cortisol in urine, plasma, and other sample matrices
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure cortisol levels down to 110 pg/ml
  • Rapid assay; get results in under four hours

Cortisol Express ELISA is a competitive assay that permits the rapid measurement of cortisol from biological samples, requiring only a two-hour incubation and one-hour development times. This ELISA offers the convenience of a fast assay while maintaining sensitivity.

Emballering: Kit contains monoclonal antibody, AChE tracer, standard, ELISA and wash buffer, polysorbate 20, coated plate, cover sheet, ellman's reagent, tracer and antiserum dye.

Advarsel: Due to an approximate 15% cross-reactivity with dexamethasone, this kit may not be suitable for use with samples that contain high concentrations of dexamethasone, such as those collected during a dexamethasone suppression test.

Specifikationer

Specifikationer

  • Pakn:
    480 Tests
  • Beskrivelse:
    Cortisol Express ELISA kit, 480 strip wells
  • Cat. No.:
    CAYM500370-480
  • Mid point:
    300 - 500 pg/ml (50% B/B₀)
  • Sensitivitet:
    110 pg/ml (80% B/B₀)
  • Detektionsområde:
    39,1 - 5000 pg/ml
  • Udviklingstid:
    120 min
  • Opbevaringstemperatur:
    –20 °C
  • Inkubationstid:
    2 hr

Specifikationer